Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ HER-2/c-erbB-2/neu Ab-1, Rabbit Polyclonal Antibody

Recommended for Immunohistochemistry (Formalin/paraffin), Western Blotting and Immunoprecipitation, Thermo Scientific™ HER-2/c-erbB-2/neu Ab-1, Rabbit Polyclonal Antibody provides accurate, reproducible results.

$272.96 - $1,223.84

Specifications

Antigen HER-2/c-erbB-2/neu Ab-1
Concentration 1mg/mL
Applications Immunoprecipitation
Applications Western Blotting
Applications Immunoprecipitation
View More Specs
Products
Catalog Number Mfr. No. Formulation Quantity Price Quantity    

RB103P

 
thermo scientific™ lab vision™
RB103P
Purified Ab with BSA and Azide 1mL Each for $1,172.94

RB103P0

 
thermo scientific™ lab vision™
RB103P0
Purified Ab with BSA and Azide 100µL Each for $272.96

RB103P1

 
thermo scientific™ lab vision™
RB103P1
Purified Ab with BSA and Azide 500µL Each for $755.61

RB103P1ABX

 
thermo scientific™ lab vision™
RB103P1ABX
Ab without BSA/Azide (Frozen) 500µL Each for $833.78

RB103PABX

 
thermo scientific™ lab vision™
RB103PABX
Ab without BSA/Azide (Frozen) 1mL Each for $1,223.84
Description & Specifications

Specifications

Antigen HER-2/c-erbB-2/neu Ab-1
Concentration 1mg/mL
Applications Immunoprecipitation
Applications Western Blotting
Applications Immunoprecipitation
Applications Western Blotting
Classification Polyclonal
Conjugate Unlabeled
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Non-Human Primate
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Non-Human Primate
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Non-Human Primate
Format Concentrated
Host Species Rabbit
Regulatory Status RUO
Research Discipline Oncogenesis
Type Primary
Immunogen Synthetic peptide derived from C-terminus of human c-erbB-2/HER-2 protein. This Sequence is identical in rat neu protein.
Isotype IgG

Recommended for Immunohistochemistry (Formalin/paraffin), Western Blotting and Immunoprecipitation, Thermo Scientific™ HER-2/c-erbB-2/neu Ab-1, Rabbit Polyclonal Antibody provides accurate, reproducible results.

c-erbB-2, second member (c-erbB-2/HER-2/neu) of the c-erbB family is a receptor tyrosine kinase. It exhibits extracellular domains with two cysteine-rich sequences, and a cytoplasmic tyrosine kinase domain flanked by large hydrophilic tails that carry several tyrosine autophosphorylation sites. Approximately 25% of primary breast and ovarian tumors were found to overexpress the protein

Host Species: Rabbit

Isotype: IgG

Species Reactivity: Human, Monkey, Rat and Mouse. Others not tested.

Epitope: C-terminal

Immunogen: Synthetic peptide derived from C-terminus of human HER-2/c-erbB-2 protein. This sequence is identical in rat neu protein.

Molecular Weight: 185 kDa

Positive Control: SKBR-3 cells, or T47D cells, or Breast CA

Cellular Localization: Membrane

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Western Blotting
  • Immunoprecipitation

Note:

USA: RUO; Int'l: RUO