Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ HER-2/c-erB-2/neu, Rabbit Monoclonal Antibody


Recommended for Immunohistochemistry (Formalin/paraffin), Thermo Scientific™ HER-2/c-erB-2/neu, Rabbit Monoclonal Antibody is part of a comprehensive antibody portfolio. Studies show rabbit monoclonal antibodies are more sensitive and specific than other available markers.

Manufacturer: thermo scientific™ lab vision™  RM9103S0

 View more versions of this product

Catalog No. RM9103S0

  • / Each
 

Description & Specifications

Specifications

Antigen HER-2/c-erB-2/neu
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone SP3
Conjugate Unlabeled
Cross Reactivity Human
Format Concentrated
Host Species Rabbit
Immunogen Recombinant protein encoding extracellular domain of human HER-2
Isotype IgG
Quantity 100μL
Regulatory Status IVD
Research Discipline Oncogenesis
Type Primary
Monoclonal or Polyclonal Monoclonal
Target Species Human

Recommended for Immunohistochemistry (Formalin/paraffin), Thermo Scientific™ HER-2/c-erB-2/neu, Rabbit Monoclonal Antibody is part of a comprehensive antibody portfolio. Studies show rabbit monoclonal antibodies are more sensitive and specific than other available markers.

c-erbB-2 is a receptor tyrosine kinase of the c-erbB family. It is closely related in structure to the epidermal growth factor receptor. c-erbB-2 oncoprotein is detectable in a proportion of breast and other adenocarconomas, as well as transitional cell carcinomas. In the case of breast cancer, expression determined by immunohistochemistry has been shown to be associated with poor prognosis.

Host Species: Rabbit

Clone: SP3

Isotype: IgG

Species Reactivity: Human. Others not tested.

Epitope: Extracellular domain

Immunogen: Recombinant protein encoding extracellular domain of human HER-2

Molecular Weight: 185 kDa

Positive Control: Breast CA

Cellular Localization: Membrane

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)