missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Heptyltriphenylphosphonium Bromide, TCI America™

$67.22 - $606.55

Chemical Identifiers

CAS 13423-48-8
Molecular Formula C25H30BrP
Molecular Weight (g/mol) 441.393
MDL Number MFCD00050249
InChI Key WCZSOHSGMBVYFW-UHFFFAOYSA-M
Synonym heptyltriphenylphosphonium bromide, n-heptyltriphenylphosphonium bromide, heptyltriphenylphosphanium bromide, heptyl triphenyl phosphanium bromide, heptyl triphenyl phosphonium bromide, n-heptyl-triphenylphosphonium bromide, 1-heptyl triphenylphosphonium bromide, phosphonium, heptyltriphenyl-, bromide, c7h15pph3br;, acmc-209bu9
PubChem CID 2724567
IUPAC Name heptyl(triphenyl)phosphanium;bromide
SMILES CCCCCCC[P+](C1=CC=CC=C1)(C2=CC=CC=C2)C3=CC=CC=C3.[Br-]
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
H0545500G View Documents TCI America
H0545500G
500g Each for $606.55
 
H054525G View Documents TCI America
H054525G
25g Each for $67.22
 
Specifications

Chemical Identifiers

13423-48-8
441.393
WCZSOHSGMBVYFW-UHFFFAOYSA-M
2724567
CCCCCCC[P+](C1=CC=CC=C1)(C2=CC=CC=C2)C3=CC=CC=C3.[Br-]
C25H30BrP
MFCD00050249
heptyltriphenylphosphonium bromide, n-heptyltriphenylphosphonium bromide, heptyltriphenylphosphanium bromide, heptyl triphenyl phosphanium bromide, heptyl triphenyl phosphonium bromide, n-heptyl-triphenylphosphonium bromide, 1-heptyl triphenylphosphonium bromide, phosphonium, heptyltriphenyl-, bromide, c7h15pph3br;, acmc-209bu9
heptyl(triphenyl)phosphanium;bromide

Specifications

White
500g
C25H30BrP
heptyltriphenylphosphonium bromide, n-heptyltriphenylphosphonium bromide, heptyltriphenylphosphanium bromide, heptyl triphenyl phosphanium bromide, heptyl triphenyl phosphonium bromide, n-heptyl-triphenylphosphonium bromide, 1-heptyl triphenylphosphonium bromide, phosphonium, heptyltriphenyl-, bromide, c7h15pph3br;, acmc-209bu9
CCCCCCC[P+](C1=CC=CC=C1)(C2=CC=CC=C2)C3=CC=CC=C3.[Br-]
441.393
441.39
≥98.0% (T)
176°C
13423-48-8
MFCD00050249
WCZSOHSGMBVYFW-UHFFFAOYSA-M
heptyl(triphenyl)phosphanium;bromide
2724567
Crystal-Powder at 20°C
Heptyltriphenylphosphonium Bromide
Safety and Handling

Safety and Handling

TSCA : No