missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Heptylphosphonic Acid, TCI America™

$235.28 - $817.38

Chemical Identifiers

CAS 4721-16-8
Molecular Formula C7H17O3P
Molecular Weight (g/mol) 180.184
MDL Number MFCD00015840
InChI Key VAJFLSRDMGNZJY-UHFFFAOYSA-N
PubChem CID 3870222
IUPAC Name heptylphosphonic acid
SMILES CCCCCCCP(=O)(O)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
H15351G View Documents TCI America
H15351G
1g Each for $235.28
 
H15355G View Documents TCI America
H15355G
5g Each for $817.38
 
Specifications

Chemical Identifiers

4721-16-8
180.184
VAJFLSRDMGNZJY-UHFFFAOYSA-N
heptylphosphonic acid
C7H17O3P
MFCD00015840
3870222
CCCCCCCP(=O)(O)O

Specifications

White-Yellow
1g
C7H17O3P
3261
CCCCCCCP(=O)(O)O
180.184
180.18
≥98.0% (T)
107°C
4721-16-8
MFCD00015840
VAJFLSRDMGNZJY-UHFFFAOYSA-N
heptylphosphonic acid
3870222
Crystal-Powder at 20°C
Heptylphosphonic Acid
Safety and Handling

Safety and Handling

EINECSNumber : (2)-1995

TSCA : No