missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Heptylbenzene 97.0+%, TCI America™
Supplier: TCI America H0046500ML
Specifications
| Heptylbenzene | |
| Colorless | |
| C13H20 | |
| 500 mL | |
| 1-phenylheptane, benzene, heptyl, n-heptylbenzene, heptane, 1-phenyl, heptane, 1 or 7-phenyl, unii-12uc21c7sn, phenylheptane, heptyl-benzene, heptane, phenyl | |
| CCCCCCCC1=CC=CC=C1 | |
| 176.303 | |
| 176.30 | |
| Liquid |
| 1078-71-3 | |
| 235°C | |
| MFCD00009545 | |
| 3082 | |
| LBNXAWYDQUGHGX-UHFFFAOYSA-N | |
| heptylbenzene | |
| 14115 | |
| ≥97.0% (GC) |
Chemical Identifiers
| 1078-71-3 | |
| 176.303 | |
| LBNXAWYDQUGHGX-UHFFFAOYSA-N | |
| 14115 | |
| CCCCCCCC1=CC=CC=C1 |
| C13H20 | |
| MFCD00009545 | |
| 1-phenylheptane, benzene, heptyl, n-heptylbenzene, heptane, 1-phenyl, heptane, 1 or 7-phenyl, unii-12uc21c7sn, phenylheptane, heptyl-benzene, heptane, phenyl | |
| heptylbenzene |
Safety and Handling
EINECSNumber : (3)-0021
TSCA : Yes