Please login to your online account to display your discounted pricing

Heptyl isocyanate, 97%, ACROS Organics™


Quantity 1g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC314340010

 View more versions of this product

Catalog No. AC314340010

  • / Each
 

Description & Specifications

Chemical Identifiers

Assay ≥96.0%
Physical Form Liquid
Formula Weight 141.22
Quantity 1g
Color Clear yellow
Density 0.87g/mL
Chemical Name or Material Heptyl isocyanate
Name Note 97%
Infrared Spectrum Authentic
Assay Percent Range 97%
Molecular Formula C8H15NO
Packaging Glass bottle
Linear Formula CH3(CH)5CHNCO
MDL Number MFCD00144892
CAS 4747-81-3
Refractive Index 1.4250 to 1.4300 (20°C, 589nm)
Health Hazard 1 GHS Signal Word: Danger
Health Hazard 2 GHS H Statement:
Causes serious eye damage.
Harmful if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF INHALED: If breathing is difficult, remove to fresh air and keep at rest in a position comfortable for breathing.
If experiencing respiratory symptoms: Call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Specifications

Chemical Name or Material Heptyl isocyanate
Name Note 97%
CAS 4747-81-3
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C8H15NO
Linear Formula CH3(CH)5CHNCO
Formula Weight 141.22
MDL Number MFCD00144892
Accessories

GHS Signal Word: Danger

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF INHALED: If breathing is difficult, remove to fresh air and keep at rest in a position comfortable for breathing.
If experiencing respiratory symptoms: Call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye damage.
Harmful if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May cause respiratory irritation.

SDS & Safety Documents