missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Heptyl beta-D-glucopyranoside, 99%

$234.41 - $422.54

Chemical Identifiers

CAS 78617-12-6
Molecular Formula C13H26O6
Molecular Weight (g/mol) 278.345
MDL Number MFCD00063299
InChI Key NIDYWHLDTIVRJT-UJPOAAIJSA-N
Synonym heptyl-beta-d-glucopyranoside, heptyl beta-d-glucopyranoside, 2r,3r,4s,5s,6r-2-heptyloxy-6-hydroxymethyl tetrahydro-2h-pyran-3,4,5-triol, b7g, n-heptyl beta-d-glucopyranoside, n-heptyl-i 2-d-glucopyranoside, heptyl-?-d-glucopyranoside, heptyl b-d-glucopyranoside, heptyl-b-d-glucopyranoside, d06ibv
PubChem CID 448173
IUPAC Name (2R,3R,4S,5S,6R)-2-heptoxy-6-(hydroxymethyl)oxane-3,4,5-triol
SMILES CCCCCCCOC1C(C(C(C(O1)CO)O)O)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAJ66576ME View Documents Alfa Aesar™
J66576ME
500mg Each for $234.41
 
AAJ6657603 View Documents Alfa Aesar™
J6657603
1g Each for $422.54
 
Specifications

Chemical Identifiers

78617-12-6
278.345
NIDYWHLDTIVRJT-UJPOAAIJSA-N
448173
CCCCCCCOC1C(C(C(C(O1)CO)O)O)O
C13H26O6
MFCD00063299
heptyl-beta-d-glucopyranoside, heptyl beta-d-glucopyranoside, 2r,3r,4s,5s,6r-2-heptyloxy-6-hydroxymethyl tetrahydro-2h-pyran-3,4,5-triol, b7g, n-heptyl beta-d-glucopyranoside, n-heptyl-i 2-d-glucopyranoside, heptyl-?-d-glucopyranoside, heptyl b-d-glucopyranoside, heptyl-b-d-glucopyranoside, d06ibv
(2R,3R,4S,5S,6R)-2-heptoxy-6-(hydroxymethyl)oxane-3,4,5-triol

Specifications

White
78617-12-6
MFCD00063299
heptyl-beta-d-glucopyranoside, heptyl beta-d-glucopyranoside, 2r,3r,4s,5s,6r-2-heptyloxy-6-hydroxymethyl tetrahydro-2h-pyran-3,4,5-triol, b7g, n-heptyl beta-d-glucopyranoside, n-heptyl-i 2-d-glucopyranoside, heptyl-?-d-glucopyranoside, heptyl b-d-glucopyranoside, heptyl-b-d-glucopyranoside, d06ibv
CCCCCCCOC1C(C(C(C(O1)CO)O)O)O
278.345
278.34
99%
500mg
C13H26O6
8806
NIDYWHLDTIVRJT-UJPOAAIJSA-N
(2R,3R,4S,5S,6R)-2-heptoxy-6-(hydroxymethyl)oxane-3,4,5-triol
448173
Powder
Heptyl beta-D-glucopyranoside
Safety and Handling

Safety and Handling

Recommended Storage : −30° to −10°C