missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Heptanophenone, 98+%, ACROS Organics™
Manufacturer: Thermo Fisher Scientific 279210250
Specifications
Heptanophenone | |
>110°C | |
Glass bottle | |
0.94 | |
Yellow | |
25g | |
1671-75-6 | |
C13H18O | |
MFCD00009539 | |
UXMQORVHJMUQFD-UHFFFAOYSA-N | |
1-phenylheptan-1-one | |
74282 | |
Liquid |
0.9400g/mL | |
Authentic | |
1.5050 to 1.508 | |
155°C (15.0mmHg) | |
17°C | |
98+% | |
98+% | |
C6H5CO(CH2)5CH3 | |
1-heptanone, 1-phenyl, 1-phenyl-1-heptanone #, 1-phenyl-heptan-1-one, enanthophenone, heptanophenone, hexyl phenyl ketone, kx07wp06jy, n-heptanophenone, unii-kx07wp06jy | |
CCCCCCC(=O)C1=CC=CC=C1 | |
190.29 | |
190.29 | |
98+% |
Chemical Identifiers
1671-75-6 | |
190.29 | |
UXMQORVHJMUQFD-UHFFFAOYSA-N | |
74282 | |
CCCCCCC(=O)C1=CC=CC=C1 |
C13H18O | |
MFCD00009539 | |
1-heptanone, 1-phenyl, 1-phenyl-1-heptanone #, 1-phenyl-heptan-1-one, enanthophenone, heptanophenone, hexyl phenyl ketone, kx07wp06jy, n-heptanophenone, unii-kx07wp06jy | |
1-phenylheptan-1-one |
Safety and Handling
EINECSNumber : 216-802-0
TSCA : TSCA