missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Heptanophenone, 98+%, ACROS Organics™

$81.64 - $81.64

Chemical Identifiers

CAS 1671-75-6
Molecular Formula C13H18O
Molecular Weight (g/mol) 190.29
MDL Number MFCD00009539
InChI Key UXMQORVHJMUQFD-UHFFFAOYSA-N
Synonym 1-heptanone, 1-phenyl
PubChem CID 74282
IUPAC Name 1-phenylheptan-1-one
SMILES CCCCCCC(=O)C1=CC=CC=C1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC279210050 SDS View Documents Acros Organics
279210050
5g Glass bottle N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC279210250 SDS View Documents Acros Organics
279210250
25g Glass bottle Each for $81.64
 
Specifications

Chemical Identifiers

1671-75-6
190.29
UXMQORVHJMUQFD-UHFFFAOYSA-N
74282
CCCCCCC(=O)C1=CC=CC=C1
C13H18O
MFCD00009539
1-heptanone, 1-phenyl
1-phenylheptan-1-one

Specifications

0.9400g/mL
Authentic
1.5050 to 1.508
155°C (15.0mmHg)
17°C
1671-75-6
C13H18O
MFCD00009539
UXMQORVHJMUQFD-UHFFFAOYSA-N
1-phenylheptan-1-one
74282
Liquid
Heptanophenone, 98+%
>110°C
Glass bottle
0.94
Yellow
5g
98+%
C6H5CO(CH2)5CH3
1-heptanone, 1-phenyl
CCCCCCC(=O)C1=CC=CC=C1
190.29
190.29
≥98.0%
Safety and Handling

Safety and Handling

EINECSNumber : 216-802-0

TSCA : TSCA