Please login to your online account to display your discounted pricing

Alfa Aesar™ Heptanophenone, 98+%

$41.14 - $156.70

Chemical Identifiers

Chemical Name or Material Heptanophenone
CAS 1671-75-6
Molecular Formula C13H18O
Formula Weight 190.29
MDL Number MFCD00009539
Beilstein 1865692
Synonym n-Hexyl phenyl ketone
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1049109

  SDS
alfa aesar™
A1049109
10g Each for $41.14

AAA1049118

  SDS
alfa aesar™
A1049118
50g Each for $156.70
Description & Specifications

Specifications

Formula Weight 190.29
Chemical Name or Material Heptanophenone
TSCA TSCA
Assay ≥98%
Molecular Formula C13H18O
MDL Number MFCD00009539
CAS 1671-75-6
Beilstein 1865692
Synonym n-Hexyl phenyl ketone
Flash Point >110°C (230°F)
Boiling Point 283°C
Melting Point ≈17°C
Quantity 10g