Please login to your online account to display your discounted pricing

Alfa Aesar™ Heptanenitrile, 98%

Chemical Identifiers

Chemical Name or Material Heptanenitrile
CAS 629-08-3
Molecular Formula CH3(CH2)5CN
Formula Weight 111.19
MDL Number MFCD00039503
UN Number UN1992
Beilstein 1740405
Synonym Enanthonitrile; Heptanonitrile
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1232606

  SDS
alfa aesar™
A1232606
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 111.19
Quantity 5g
UN Number UN1992
Melting Point −64°C
Chemical Name or Material Heptanenitrile
Assay 98%
CAS 629-08-3
Synonym Enanthonitrile; Heptanonitrile
Health Hazard 1 Danger
Boiling Point 186° to 187°C
Beilstein 1740405
Health Hazard 2 GHS H Statement
H331-H226-H302-H312
Toxic if inhaled.
Flammable liquid and vapor.
Harmful if swallowed.
Harmful in contact with skin.
Flash Point 58°C (136°F)
TSCA TSCA
DOT Information Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, TOXIC, N.O.S.
Health Hazard 3 GHS P Statement
P210-P261-P280-P303+P361+P353-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
MDL Number MFCD00039503
Molecular Formula CH3(CH2)5CN

Applications
The Heptanenitrile is used for producing 2, 3, 5, 6-tetrahexyl-pyrazine. As intermediates.

Solubility
Slightly soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, Strong reducing agents, Strong acids, Strong bases.

Safety and Handling

Danger

GHS P Statement
P210-P261-P280-P303+P361+P353-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H331-H226-H302-H312
Toxic if inhaled.
Flammable liquid and vapor.
Harmful if swallowed.
Harmful in contact with skin.