missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Heptanenitrile, 98%

Manufacturer:  Alfa Aesar™ A1232614

 View more versions of this product

Catalog No. AAA1232614



Description

Description

Applications
The Heptanenitrile is used for producing 2, 3, 5, 6-tetrahexyl-pyrazine. As intermediates.

Solubility
Slightly soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, Strong reducing agents, Strong acids, Strong bases.
Specifications

Specifications

Heptanenitrile
186°C to 187°C
25g
C7H13N
UN1992
heptanonitrile, hexyl cyanide, enanthonitrile, 1-cyanohexane, 1-heptanonitrile, heptane-1-nitrile, n-heptanenitrile, unii-ua4z3qt98a, ua4z3qt98a, heptane nitrile
CCCCCCC#N
111.188
111.19
58°C (136°F)
-64°C
629-08-3
MFCD00039503
1740405
SDAXRHHPNYTELL-UHFFFAOYSA-N
heptanenitrile
12372
98%

Chemical Identifiers

629-08-3
111.188
SDAXRHHPNYTELL-UHFFFAOYSA-N
12372
CCCCCCC#N
C7H13N
MFCD00039503
heptanonitrile, hexyl cyanide, enanthonitrile, 1-cyanohexane, 1-heptanonitrile, heptane-1-nitrile, n-heptanenitrile, unii-ua4z3qt98a, ua4z3qt98a, heptane nitrile
heptanenitrile
Safety and Handling

Safety and Handling

GHS H Statement
H331-H226-H302-H312
Toxic if inhaled.
Flammable liquid and vapor.
Harmful if swallowed.
Harmful in contact with skin.

GHS P Statement
P210-P261-P280-P303+P361+P353-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, TOXIC, N.O.S.

TSCA : TSCA

SDS Documents