Please login to your online account to display your discounted pricing

Heptane OmniSolv™, EMD Millipore

Low UV Cutoff

$261.40 - $261.40

Chemical Identifiers

Chemical Name or Material Heptane
CAS 142-82-5
Molecular Formula C7H16
Formula Weight 100.2
UN Number 1206
Synonym n-Heptane
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

MHX00761

 
emd millipore corporation
HX00761
Glass Bottle 1L Each for $261.40
Description & Specifications

Specifications

Assay ≥99.0%
Melting Point −91.11°C
Formula Weight 100.2
Boiling Point 98.34°C
Quantity 1L
Storage Information Store in accordance with local regulations. Store in a segregated and approved area. Store in original container, protected from direct sunlight. Eliminate all ignition sources. Separate from oxidizing materials. Keep container tightly closed and sealed until ready for use. Containers that have been opened must be carefully resealed and kept upright to prevent leakage.
Molecular Formula C7H16
Chemical Name or Material Heptane
Synonym n-Heptane
Physical Form Liquid
Density 0.68g/cm3 at 20°C
Color Colorless
Ignition Point 221.85°C (431.3°F)
Odor Charactristic
Packaging Glass Bottle
Flash Point −4.15°C (−24.5°F), Closed cup
CAS 142-82-5
UN Number 1206