missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Heptane (HPLC), Fisher Chemical

Manufacturer:  Fisher Chemical H350SK1

 View more versions of this product

Catalog No. H350SK-1




Specifications

Specifications

Heptane
142-82-5,28729-52-4,26635-64-3,108-87-2
Mobile phase for HPLC applications
C7H16
MFCD00009544
dipropyl methane, dipropylmethane, eptani, gettysolve-c, heptan, heptan polish, heptanen, heptyl hydride, n-heptane, skellysolve c
CCCCCCC
100.205
CHEBI:43098
Liquid
HPLC
Pass Test
Safe-Cote™ Amber Glass 
5ppm max.
0.02% max.
0.4 mPa/s at 20°C
Colorless
1L
0.014 AU max. at 254nm
Vapor Pressure: 48mbar at 20°C
96% min by GC
CH3(CH2)5CH3
1206
IMNFDUFMRHMDMM-UHFFFAOYSA-N
heptane
8900
100.2
≥96%
0.683g/cm³
Pass Test
1.3845 to 1.3865 (at 25°C)
48mbar at 20°C
-91°C (195.8°F)
98°C
-91°C

Chemical Identifiers

142-82-5
100.205
IMNFDUFMRHMDMM-UHFFFAOYSA-N
8900
heptane
C7H16
MFCD00009544
dipropyl methane, dipropylmethane, eptani, gettysolve-c, heptan, heptan polish, heptanen, heptyl hydride, n-heptane, skellysolve c
CHEBI:43098
CCCCCCC
Safety and Handling

Safety and Handling

DANGER!

Emergency Overview
Flammable liquid and vapor. Inhalation may cause central nervous system effects. May cause irritation of respiratory tract. Aspiration hazard if swallowed - can enter lungs and cause damage. Very toxic to aquatic organisms, may cause long-term adverse effects in the aquatic environment. Use personal protective equipment. Keep away from open flames, hot surfaces and sources of ignition. Use only under a chemical fume hood. Wash off immediately with plenty of water for at least 15 minutes. Obtain medical attention. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Obtain medical attention. Move to fresh air.

NFPA

Health:1
Flammability:3
Instability:0

DANGER!

DOTInformation : DOT Class 3, : Flammable Liquid

SDS Documents