Please login to your online account to display your discounted pricing

Heptane ChromAR, Macron Fine Chemicals™

macron

Chemical Identifiers

Chemical Name or Material Heptane
CAS 142-82-5
Molecular Formula CH3(CH2)5CH3
Formula Weight 100.2
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

MK51391

 
avantor performance
513906
Glass 1L CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

MK51394

 
avantor performance
513910
Glass 4L CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 100.2
Packaging Glass
CAS 142-82-5
Molecular Formula CH3(CH2)5CH3
Chemical Name or Material Heptane
Quantity 1L