missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Heptane, 99.0% min., EMD Millipore

Manufacturer:  Millipore Sigma HX008020

 View more versions of this product

Catalog No. MHX008020



Specifications

Specifications

Heptane
99.0% min.
n-heptane, dipropylmethane, heptan, heptyl hydride, dipropyl methane, gettysolve-c, skellysolve c, heptanen, eptani, heptan polish
CCCCCCC
100.205
CHEBI:43098
Liquid
200L
142-82-5
C7H16
IMNFDUFMRHMDMM-UHFFFAOYSA-N
heptane
8900
100.21
Colorless

Chemical Identifiers

142-82-5
100.205
n-heptane, dipropylmethane, heptan, heptyl hydride, dipropyl methane, gettysolve-c, skellysolve c, heptanen, eptani, heptan polish
CHEBI:43098
CCCCCCC
C7H16
IMNFDUFMRHMDMM-UHFFFAOYSA-N
8900
heptane
Documents