missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Heptane, 99.0% min., EMD Millipore

$107.89 - $9,480.91

Chemical Identifiers

CAS 142-82-5
Physical Form Liquid
Assay Percent Range 99.0% min.
Molecular Formula CH3(CH2)5CH3
Formula Weight 100.21
Synonym n-Heptane
Chemical Name or Material Heptane
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
MHX008020 Emd Millipore Corporation
HX008020
200L Each for $9,480.91
MHX00806 Emd Millipore Corporation
HX00806
500mL Each for $107.89
Specifications Description & Specifications

Chemical Identifiers

142-82-5
99.0% min.
100.21
Heptane
Liquid
CH3(CH2)5CH3
n-Heptane

Specifications

100.21
Clear colorless
CH3(CH2)5CH3
99.0% min.
Heptane
Liquid
200L
142-82-5
n-Heptane