Please login to your online account to display your discounted pricing

Alfa Aesar™ Heptanal dimethyl acetal, 96%

$52.49 - $155.47

Chemical Identifiers

Chemical Name or Material Heptanal dimethyl acetal
CAS 10032-05-0
Molecular Formula CH3(CH2)5CH(OCH3)2
Formula Weight 160.26
MDL Number MFCD00014908
Beilstein 1698661
Synonym 1,1-Dimethoxyheptane; Heptaldehyde dimethyl acetal
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0433514

  SDS
alfa aesar™
L0433514
25g Each for $52.49

AAL0433522

  SDS
alfa aesar™
L0433522
100g Each for $155.47
Description & Specifications

Specifications

Formula Weight 160.26
Chemical Name or Material Heptanal dimethyl acetal
TSCA TSCA
Assay 96%
Molecular Formula CH3(CH2)5CH(OCH3)2
MDL Number MFCD00014908
CAS 10032-05-0
Synonym 1,1-Dimethoxyheptane; Heptaldehyde dimethyl acetal
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H227
Combustible liquid.
Health Hazard 3 GHS P Statement
P210-P280-P370+P378a-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
In case of fire: Use for extinction: CO2, powder or water spray.
Store in a well-ventilated place. Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Quantity 25g
Beilstein 1698661
Flash Point 68°C (154°F)
Boiling Point 60°C (1mmHg)
Safety and Handling

Warning

GHS P Statement
P210-P280-P370+P378a-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
In case of fire: Use for extinction: CO2, powder or water spray.
Store in a well-ventilated place. Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H227
Combustible liquid.