missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Heptanal dimethyl acetal, 96%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals L0433514
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Heptanal dimethyl acetal | |
0.849 | |
68°C (154°F) | |
1.4115 | |
25 g | |
heptanal dimethyl acetal, heptane, 1,1-dimethoxy, enanthal dimethyl acetal, oenanthal dimethyl acetal, heptanal, dimethyl acetal, heptaldehyde dimethyl acetal, aldehyde c-7 dimethyl acetal, unii-rmc47kwz4j, fema no. 2541, 1,1-dimethoxy-heptane | |
CCCCCCC(OC)OC | |
160.26 | |
160.26 |
10032-05-0 | |
60°C (1 mmHg) | |
C9H20O2 | |
MFCD00014908 | |
1698661 | |
BBMCNYFBAIUERL-UHFFFAOYSA-N | |
1,1-dimethoxyheptane | |
61453 | |
96% |
Chemical Identifiers
10032-05-0 | |
160.26 | |
BBMCNYFBAIUERL-UHFFFAOYSA-N | |
61453 | |
CCCCCCC(OC)OC |
C9H20O2 | |
MFCD00014908 | |
heptanal dimethyl acetal, heptane, 1,1-dimethoxy, enanthal dimethyl acetal, oenanthal dimethyl acetal, heptanal, dimethyl acetal, heptaldehyde dimethyl acetal, aldehyde c-7 dimethyl acetal, unii-rmc47kwz4j, fema no. 2541, 1,1-dimethoxy-heptane | |
1,1-dimethoxyheptane |
Safety and Handling
GHS H Statement
H227
Combustible liquid.
P210-P280-P370+P378q-P501c
H227
EINECSNumber : 233-103-6
TSCA : Yes
Recommended Storage : Ambient temperatures