Learn More
Heptanal, 97%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals B23830AU
Description
Heptanal is used as a synthesis intermediate in the fragrances and flavors industry. It is the precursor to 1-heptanol, ethyl heptanoate and for certain lubricants.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsHeptanal is used as a synthesis intermediate in the fragrances and flavors industry. It is the precursor to 1-heptanol, ethyl heptanoate and for certain lubricants.
Solubility
Soluble in water 1.3 g/L.
Notes
Hygroscopic. Store away from oxidizing agents, air.
Specifications
Heptanal | |
-43°C | |
152°C to 154°C | |
0.97 | |
C7H14O | |
MFCD00007028 | |
UN3056 | |
Air Sensitive | |
heptaldehyde, enanthaldehyde, n-heptaldehyde, enanthal, heptyl aldehyde, heptanaldehyde, n-heptanal, oenanthaldehyde, enanthole, oenanthal | |
FXHGMKSSBGDXIY-UHFFFAOYSA-N | |
heptanal | |
8130 | |
114.19 | |
Liquid |
111-71-7 | |
0.85 | |
35°C (95°F) | |
Unpleasant | |
1.4125 | |
1000 mL | |
1560236 | |
14,4658 | |
Soluble in water 1.3g/L. | |
CCCCCCC=O | |
114.19 | |
CHEBI:34787 | |
97% |
Chemical Identifiers
111-71-7 | |
114.19 | |
FXHGMKSSBGDXIY-UHFFFAOYSA-N | |
8130 | |
heptanal |
C7H14O | |
MFCD00007028 | |
heptaldehyde, enanthaldehyde, n-heptaldehyde, enanthal, heptyl aldehyde, heptanaldehyde, n-heptanal, oenanthaldehyde, enanthole, oenanthal | |
CHEBI:34787 | |
CCCCCCC=O |
Safety and Handling
GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapour.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P210-P233-P240-P241-P242-P243-P264b-P280-P303+P361+P353-P305+P351+P338-P332+P313-P363-P370+P378q-P501c
H226-H315
DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: N-HEPTALDEHYDE
EINECSNumber : 203-898-4
RTECSNumber : MI6900000
TSCA : Yes
storageNote1 : Hygroscopic
Recommended Storage : Ambient temperatures
RUO – Research Use Only