missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Heptamethyleneimine, TCI America™

$261.25 - $883.08

Chemical Identifiers

CAS 1121-92-2
Molecular Formula C7H15N
Molecular Weight (g/mol) 113.204
MDL Number MFCD00003270
InChI Key QXNDZONIWRINJR-UHFFFAOYSA-N
Synonym Azacyclooctane
PubChem CID 14276
ChEBI CHEBI:38792
IUPAC Name azocane
SMILES C1CCCNCCC1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
H05465G View Documents TCI America
H05465G
5g Each for $261.25
 
H054625G View Documents TCI America
H054625G
25g Each for $883.08
 
Specifications

Chemical Identifiers

1121-92-2
113.204
QXNDZONIWRINJR-UHFFFAOYSA-N
14276
azocane
C7H15N
MFCD00003270
Azacyclooctane
CHEBI:38792
C1CCCNCCC1

Specifications

162°C
5g
C7H15N
2920
QXNDZONIWRINJR-UHFFFAOYSA-N
azocane
14276
113.20
≥98.0% (GC)
Yellow
1121-92-2
MFCD00003270
Azacyclooctane
C1CCCNCCC1
113.204
CHEBI:38792
Clear Liquid at 20°C
Heptamethyleneimine
Safety and Handling

Safety and Handling

TSCA : Yes