Please login to your online account to display your discounted pricing

Heptakis-6-iodo-6-deoxy-β-cyclodextrin, ACROS Organics™

Chemical Identifiers

Chemical Name or Material Heptakis-6-iodo-6-deoxy-β-cyclodextrin
CAS 30754-23-5
Molecular Formula C42H63I7O28
Formula Weight 1904.28
MDL Number MFCD00803290
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC299011000

  SDS
thermo fisher scientific
299011000
Glass bottle 100mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC299015000

  SDS
thermo fisher scientific
299015000
Glass bottle 500mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form Solid
Formula Weight 1904.28
Melting Point 218° to 222°C (dec.)
Quantity 100mg
Color Off-white
NMR conform
Packaging Glass bottle
Molecular Formula C42H63I7O28
MDL Number MFCD00803290
Chemical Name or Material Heptakis-6-iodo-6-deoxy-β-cyclodextrin
CAS 30754-23-5

Chelation/Complexation reagent