missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Heptafluoropropyl Iodide (stabilized with Copper chip), TCI America™

$67.22 - $224.59

Chemical Identifiers

CAS 754-34-7
Molecular Formula C3F7I
Molecular Weight (g/mol) 295.926
MDL Number MFCD00001061
InChI Key XTGYEAXBNRVNQU-UHFFFAOYSA-N
Synonym heptafluoro-1-iodopropane, heptafluoropropyl iodide, perfluoropropyl iodide, 1-iodoheptafluoropropane, 1-iodoperfluoropropane, propane, heptafluoro-1-iodo, 1-perfluoropropyl iodide, heptafluoroiodopropane, iodoheptafluoropropane, propane, heptafluoroiodo
PubChem CID 33977
IUPAC Name 1,1,1,2,2,3,3-heptafluoro-3-iodopropane
SMILES C(C(F)(F)F)(C(F)(F)I)(F)F
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
H05965G View Documents TCI America
H05965G
5g Each for $67.22
 
H059625G View Documents TCI America
H059625G
25g Each for $224.59
 
Specifications

Chemical Identifiers

754-34-7
295.926
XTGYEAXBNRVNQU-UHFFFAOYSA-N
33977
C(C(F)(F)F)(C(F)(F)I)(F)F
C3F7I
MFCD00001061
heptafluoro-1-iodopropane, heptafluoropropyl iodide, perfluoropropyl iodide, 1-iodoheptafluoropropane, 1-iodoperfluoropropane, propane, heptafluoro-1-iodo, 1-perfluoropropyl iodide, heptafluoroiodopropane, iodoheptafluoropropane, propane, heptafluoroiodo
1,1,1,2,2,3,3-heptafluoro-3-iodopropane

Specifications

41°C
-95°C
754-34-7
MFCD00001061
XTGYEAXBNRVNQU-UHFFFAOYSA-N
1,1,1,2,2,3,3-heptafluoro-3-iodopropane
33977
Clear Liquid at 20°C
Heptafluoropropyl Iodide (stabilized with Copper chip)
Red
5g
C3F7I
heptafluoro-1-iodopropane, heptafluoropropyl iodide, perfluoropropyl iodide, 1-iodoheptafluoropropane, 1-iodoperfluoropropane, propane, heptafluoro-1-iodo, 1-perfluoropropyl iodide, heptafluoroiodopropane, iodoheptafluoropropane, propane, heptafluoroiodo
C(C(F)(F)F)(C(F)(F)I)(F)F
295.926
295.93
≥95.0% (GC)
Safety and Handling

Safety and Handling

RTECSNumber : TZ3930000

TSCA : Yes