missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Heptafluoroisopropyl Iodide 95.0+%, TCI America™
$121.11 - $353.37
Chemical Identifiers
CAS | 677-69-0 |
---|---|
Molecular Formula | C3F7I |
Molecular Weight (g/mol) | 295.926 |
MDL Number | MFCD00042089 |
InChI Key | BBZVTTKMXRPMHZ-UHFFFAOYSA-N |
Synonym | heptafluoroisopropyl iodide, perfluoroisopropyl iodide, heptafluoro-2-iodopropane, 2-iodoheptafluoropropane, propane, 1,1,1,2,3,3,3-heptafluoro-2-iodo, 2-iodoperfluoropropane, propane, heptafluoro-2-iodo, perfluoro-2-iodopropane, heptafluoroisopropyliodide, 2-heptafluoroisopropyliodide |
PubChem CID | 33978 |
IUPAC Name | 1,1,1,2,3,3,3-heptafluoro-2-iodopropane |
SMILES | C(C(F)(F)F)(C(F)(F)F)(F)I |
Chemical Identifiers
677-69-0 | |
295.926 | |
BBZVTTKMXRPMHZ-UHFFFAOYSA-N | |
33978 | |
C(C(F)(F)F)(C(F)(F)F)(F)I |
C3F7I | |
MFCD00042089 | |
heptafluoroisopropyl iodide, perfluoroisopropyl iodide, heptafluoro-2-iodopropane, 2-iodoheptafluoropropane, propane, 1,1,1,2,3,3,3-heptafluoro-2-iodo, 2-iodoperfluoropropane, propane, heptafluoro-2-iodo, perfluoro-2-iodopropane, heptafluoroisopropyliodide, 2-heptafluoroisopropyliodide | |
1,1,1,2,3,3,3-heptafluoro-2-iodopropane |
Specifications
677-69-0 | |
40°C | |
MFCD00042089 | |
heptafluoroisopropyl iodide, perfluoroisopropyl iodide, heptafluoro-2-iodopropane, 2-iodoheptafluoropropane, propane, 1,1,1,2,3,3,3-heptafluoro-2-iodo, 2-iodoperfluoropropane, propane, heptafluoro-2-iodo, perfluoro-2-iodopropane, heptafluoroisopropyliodide, 2-heptafluoroisopropyliodide | |
C(C(F)(F)F)(C(F)(F)F)(F)I | |
295.926 | |
295.93 | |
Liquid |
Red | |
C3F7I | |
25 g | |
BBZVTTKMXRPMHZ-UHFFFAOYSA-N | |
1,1,1,2,3,3,3-heptafluoro-2-iodopropane | |
33978 | |
≥95.0% (GC) | |
Heptafluoroisopropyl Iodide |
Safety and Handling
TSCA : Yes
Recommended Storage : Refrigerator