Please login to your online account to display your discounted pricing

Heptafluorobutyryl chloride, 98%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material Heptafluorobutyryl chloride
Name Note 98%
CAS 375-16-6
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C4ClF7O
Linear Formula CF3CFCFCOCl
Formula Weight 232.49
MDL Number MFCD00000707
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC312100250

  SDS
thermo fisher scientific
312100250
Glass bottle 25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥97.5%
Physical Form Liquid
Formula Weight 232.49
Quantity 25g
Color Clear colorless
Beilstein 02, IV, 818
EINECS Number 206-785-8
TSCA TSCA
Density 1.55g/mL
Refractive Index 1.2860 to 1.2940 (20°C, 589nm)
Chemical Name or Material Heptafluorobutyryl chloride
Infrared Spectrum Authentic
Packaging Glass bottle
Name Note 98%
Assay Percent Range 98%
Molecular Formula C4ClF7O
Linear Formula CF3CFCFCOCl
MDL Number MFCD00000707
CAS 375-16-6
Health Hazard 1 GHS Signal Word: Danger
Health Hazard 2 GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.
Health Hazard 3 GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.