Please login to your online account to display your discounted pricing

Heptafluorobutyric Acid, Sequenation Grade 99%, ACROS Organics™


Quantity 150mL
Packaging Glass bottle

Manufacturer: thermo fisher scientific  41122-1500

 View more versions of this product

Catalog No. AC411221500

  • / Each
 

Description & Specifications

Chemical Identifiers

Assay ≥98.5% (Acid)
Physical Form Powder
Formula Weight 214.04
Color Beige to orange-brown
Quantity 150mL
Water 0.1% Max. (K.F.)
EINECS Number 206-786-3
TSCA TSCA
Density 1.6400g/mL
RTECS Number ET4025000
Absorbance (0.1% aqueous solution)
Health Hazard 2 GHS P Statement Causes severe skin burns and eye damage.
Chemical Name or Material Heptafluorobutyric acid
Infrared Spectrum Authentic
Packaging Glass bottle
Assay Percent Range 99%
Molecular Formula C4HF7O2
Linear Formula CF3CF2CF2CO2H
MDL Number MFCD00004171
CAS 375-22-4
Synonym HFBA
Absorbance Conditions (0.1% aqueous solution)
Health Hazard 1 Danger
Health Hazard 3 GHS P Statement IF SWALLOWED: rinse mouth. Do NOT induce vomiting. Wear eye protection/face protection. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Co

Specifications

Chemical Name or Material Heptafluorobutyric acid
Assay ≥98.5% (Acid)
Assay Percent Range 99%
Molecular Formula C4HF7O2
Linear Formula CF3CF2CF2CO2H
Formula Weight 214.04
MDL Number MFCD00004171
Synonym HFBA
Accessories

Danger

GHS P Statement IF SWALLOWED: rinse mouth. Do NOT induce vomiting. Wear eye protection/face protection. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Co

GHS P Statement Causes severe skin burns and eye damage.

SDS & Safety Documents