missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Heptafluorobutyric Acid, Sequenation Grade 99%, ACROS Organics™

$1,283.92 - $1,283.92

Chemical Identifiers

Assay Percent Range 99%
Molecular Formula C4HF7O2
Linear Formula CF3CF2CF2CO2H
Formula Weight 214.04
MDL Number MFCD00004171
Synonym HFBA
Chemical Name or Material Heptafluorobutyric acid
Assay ≥98.5% (Acid)
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
AC411221500 SDS Thermo Fisher Scientific
41122-1500
Glass bottle 150mL Each for $1,283.92
Specifications Description & Specifications

Chemical Identifiers

99%
CF3CF2CF2CO2H
MFCD00004171
Heptafluorobutyric acid
C4HF7O2
214.04
HFBA
≥98.5% (Acid)

Specifications

(0.1% aqueous solution)
150mL
(0.1% aqueous solution)
Authentic
Powder
C4HF7O2
214.04
HFBA
Heptafluorobutyric acid
Beige to orange-brown
375-22-4
1.6400g/mL
Glass bottle
99%
CF3CF2CF2CO2H
MFCD00004171
0.1% Max. (K.F.)
≥98.5% (Acid)

Safety and Handling

GHS H Statement
Causes severe skin burns and eye damage. May be corrosive to metals.

GHS P Statement
IF SWALLOWED: rinse mouth. Do NOT induce vomiting. Wear protective gloves/protective clothing/eye protection/face protection. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Immediately call a POISON CENTER or doctor/physician. IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/shower.

Danger

EINECSNumber : 206-786-3

RTECSNumber : ET4025000

TSCA : TSCA