Please login to your online account to display your discounted pricing

Alfa Aesar™ Heptafluorobutyramide, 97%

$76.05 - $281.69

Chemical Identifiers

Chemical Name or Material Heptafluorobutyramide
CAS 662-50-0
Molecular Formula CF3CF2CF2CONH2
Formula Weight 213.06
MDL Number MFCD00039773
Beilstein 1790527
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2146618

 
alfa aesar™
B2146618
50g Each for $281.69

AAB2146609

 
alfa aesar™
B2146609
10g Each for $76.05
Description & Specifications

Specifications

Formula Weight 213.06
Chemical Name or Material Heptafluorobutyramide
TSCA TSCA
Assay 97%
Molecular Formula CF3CF2CF2CONH2
MDL Number MFCD00039773
CAS 662-50-0
Beilstein 1790527
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
RTECS Number ES4725000
Melting Point 102° to 106°C
Quantity 50g
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.