missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Heptafluoro-2-iodopropane, 97%, stab. with copper powder

$50.12 - $419.46

Chemical Identifiers

CAS 677-69-0
Molecular Formula C3F7I
Molecular Weight (g/mol) 295.926
MDL Number MFCD00042089
InChI Key BBZVTTKMXRPMHZ-UHFFFAOYSA-N
Synonym heptafluoroisopropyl iodide, perfluoroisopropyl iodide, heptafluoro-2-iodopropane, 2-iodoheptafluoropropane, propane, 1,1,1,2,3,3,3-heptafluoro-2-iodo, 2-iodoperfluoropropane, propane, heptafluoro-2-iodo, perfluoro-2-iodopropane, heptafluoroisopropyliodide, 2-heptafluoroisopropyliodide
PubChem CID 33978
IUPAC Name 1,1,1,2,3,3,3-heptafluoro-2-iodopropane
SMILES C(C(F)(F)F)(C(F)(F)F)(F)I
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL1652306 View Documents Alfa Aesar™
L1652306
5g Each for $50.12
 
AAL1652314 View Documents Alfa Aesar™
L1652314
25g Each for $144.65
 
AAL1652322 View Documents Alfa Aesar™
L1652322
100g Each for $419.46
 
Specifications

Chemical Identifiers

677-69-0
295.926
BBZVTTKMXRPMHZ-UHFFFAOYSA-N
33978
C(C(F)(F)F)(C(F)(F)F)(F)I
C3F7I
MFCD00042089
heptafluoroisopropyl iodide, perfluoroisopropyl iodide, heptafluoro-2-iodopropane, 2-iodoheptafluoropropane, propane, 1,1,1,2,3,3,3-heptafluoro-2-iodo, 2-iodoperfluoropropane, propane, heptafluoro-2-iodo, perfluoro-2-iodopropane, heptafluoroisopropyliodide, 2-heptafluoroisopropyliodide
1,1,1,2,3,3,3-heptafluoro-2-iodopropane

Specifications

38°C
677-69-0
C3F7I
MFCD00042089
heptafluoroisopropyl iodide, perfluoroisopropyl iodide, heptafluoro-2-iodopropane, 2-iodoheptafluoropropane, propane, 1,1,1,2,3,3,3-heptafluoro-2-iodo, 2-iodoperfluoropropane, propane, heptafluoro-2-iodo, perfluoro-2-iodopropane, heptafluoroisopropyliodide, 2-heptafluoroisopropyliodide
C(C(F)(F)F)(C(F)(F)F)(F)I
295.926
295.93
Heptafluoro-2-iodopropane, stabilized with copper powder
-56°C
5g
(CF3)2CFI
1841228
BBZVTTKMXRPMHZ-UHFFFAOYSA-N
1,1,1,2,3,3,3-heptafluoro-2-iodopropane
33978
97%
Safety and Handling

Safety and Handling

GHS H Statement
H332-H315-H319-H335
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

RTECSNumber : TZ3925000

TSCA : TSCA

Recommended Storage : 2° to 8°C