Please login to your online account to display your discounted pricing

Alfa Aesar™ Heptafluoro-1-iodopropane, 97%, stab. with copper

$76.54 - $76.54

Chemical Identifiers

Chemical Name or Material Heptafluoro-1-iodopropane
CAS 754-34-7
Name Note stabilized with copper
Molecular Formula CF3CF2CF2I
Formula Weight 295.93
MDL Number MFCD00001061
Beilstein 1758822
Synonym Heptafluoro-n-propyl iodide; Perfluoropropyl iodide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1478406

  SDS
alfa aesar™
A1478406
5g Each for $76.54
Description & Specifications

Specifications

Formula Weight 295.93
Chemical Name or Material Heptafluoro-1-iodopropane
Boiling Point 40° to 41°C
TSCA TSCA
Name Note stabilized with copper
Melting Point −95°C
Assay 97%
Molecular Formula CF3CF2CF2I
MDL Number MFCD00001061
CAS 754-34-7
Quantity 5g
Beilstein 1758822
Synonym Heptafluoro-n-propyl iodide; Perfluoropropyl iodide
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Recommended Storage 2° to 8°C
RTECS Number TZ3930000
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

2° to 8°C