missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Heptafluoro-1-iodopropane, 97%, stab. with copper

Manufacturer:  Alfa Aesar™ A1478414

 View more versions of this product

Catalog No. AAA1478414



Specifications

Specifications

Heptafluoro-1-iodopropane
stabilized with copper
754-34-7
C3F7I
1758822
XTGYEAXBNRVNQU-UHFFFAOYSA-N
1,1,1,2,2,3,3-heptafluoro-3-iodopropane
33977
97%
40°C to 41°C
-95°C
25g
MFCD00001061
heptafluoro-1-iodopropane, heptafluoropropyl iodide, perfluoropropyl iodide, 1-iodoheptafluoropropane, 1-iodoperfluoropropane, propane, heptafluoro-1-iodo, 1-perfluoropropyl iodide, heptafluoroiodopropane, iodoheptafluoropropane, propane, heptafluoroiodo
C(C(F)(F)F)(C(F)(F)I)(F)F
295.926
295.93

Chemical Identifiers

754-34-7
295.926
XTGYEAXBNRVNQU-UHFFFAOYSA-N
33977
C(C(F)(F)F)(C(F)(F)I)(F)F
C3F7I
MFCD00001061
heptafluoro-1-iodopropane, heptafluoropropyl iodide, perfluoropropyl iodide, 1-iodoheptafluoropropane, 1-iodoperfluoropropane, propane, heptafluoro-1-iodo, 1-perfluoropropyl iodide, heptafluoroiodopropane, iodoheptafluoropropane, propane, heptafluoroiodo
1,1,1,2,2,3,3-heptafluoro-3-iodopropane
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

RTECSNumber : TZ3930000

TSCA : TSCA

Recommended Storage : 2° to 8°C

SDS Documents