missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ HEPPSO, 0.2M buffer soln., pH 7.0

$54.59 - $93.92

Chemical Identifiers

CAS 68399-78-0
Molecular Formula C9H20N2O5S
Molecular Weight (g/mol) 268.328
MDL Number MFCD00038351
InChI Key GIZQLVPDAOBAFN-UHFFFAOYSA-N
PubChem CID 100205
ChEBI CHEBI:32951
IUPAC Name 2-hydroxy-3-[4-(2-hydroxyethyl)piperazin-1-yl]propane-1-sulfonic acid
SMILES C1CN(CCN1CCO)CC(CS(=O)(=O)O)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAJ63877AE View Documents Alfa Aesar™
J63877AE
100mL Each for $54.59
 
AAJ63877AK View Documents Alfa Aesar™
J63877AK
250mL Each for $93.92
 
Specifications

Chemical Identifiers

68399-78-0
268.328
GIZQLVPDAOBAFN-UHFFFAOYSA-N
CHEBI:32951
C1CN(CCN1CCO)CC(CS(=O)(=O)O)O
C9H20N2O5S
MFCD00038351
100205
2-hydroxy-3-[4-(2-hydroxyethyl)piperazin-1-yl]propane-1-sulfonic acid

Specifications

68399-78-0
MFCD00038351
GIZQLVPDAOBAFN-UHFFFAOYSA-N
2-hydroxy-3-[4-(2-hydroxyethyl)piperazin-1-yl]propane-1-sulfonic acid
100205
Liquid
Colorless
HEPPSO buffer soln.
C9H20N2O5S
0.2M buffer soln.
C1CN(CCN1CCO)CC(CS(=O)(=O)O)O
268.328
CHEBI:32951
7.0
100mL
Safety and Handling

Safety and Handling

TSCA : TSCA

Recommended Storage : 2° to 8°C