Please login to your online account to display your discounted pricing

HEPES Hemisodium Salt, MP Biomedicals™


Quantity 25g

Manufacturer: mp biomedicals  0215245125

 View more versions of this product

Catalog No. ICN15245125

  • / Each
 

Description & Specifications

Chemical Identifiers

Quantity 25g
Chemical Name or Material HEPES Hemisodium Salt
CAS 103404-87-1
Physical Form White powder
Assay Percent Range 1.005
Molecular Formula C8H17N2O4S· 1/2Na
Formula Weight 249.3g/mol
MDL Number MFCD00036463
Synonym 4-(2-Hydroxyethyl)piperazine-1-ethanesulfonic acid hemisodium salt
Recommended Storage Store at Room Temperature (15 to 30°C). Store Desiccated.

Specifications

Chemical Name or Material HEPES Hemisodium Salt
CAS 103404-87-1
Physical Form White powder
Assay Percent Range 1.005
Molecular Formula C8H17N2O4S· 1/2Na
Formula Weight 249.3g/mol
MDL Number MFCD00036463
Synonym 4-(2-Hydroxyethyl)piperazine-1-ethanesulfonic acid hemisodium salt
Accessories

Store at Room Temperature (15 to 30°C). Store Desiccated.