Please login to your online account to display your discounted pricing

Alfa Aesar™ HEPES-buffered saline (5X, pH 7.4), low endotoxin

Chemical Identifiers

Chemical Name or Material HEPES-buffered saline (5X, pH 7.4)
Name Note low endotoxin
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAJ67502AE

  SDS
alfa aesar™
J67502AE
100mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAJ67502AK

  SDS
alfa aesar™
J67502AK
250mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Chemical Name or Material HEPES-buffered saline (5X, pH 7.4)
Color Colorless
Quantity 100mL
TSCA TSCA
Name Note low endotoxin
Physical Form Liquid
Storage Note 1 2° to 8°C
Safety and Handling

2° to 8°C