missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ HEPES Buffer, 0.2M buffer soln., pH 8.0, low endotoxin

Manufacturer:  Alfa Aesar™ J67470AK

 View more versions of this product

Catalog No. AAJ67470AK



Specifications

Specifications

HEPES Buffer
7365-45-9
MFCD00006158
JKMHFZQWWAIEOD-UHFFFAOYSA-N
2-[4-(2-hydroxyethyl)piperazin-1-yl]ethanesulfonic acid
23831
Liquid
Colorless
low endotoxin
C8H18N2O4S
0.2M buffer soln.
C1CN(CCN1CCO)CCS(=O)(=O)O
238.302
CHEBI:42334
8.0
250mL

Chemical Identifiers

7365-45-9
238.302
JKMHFZQWWAIEOD-UHFFFAOYSA-N
CHEBI:42334
C1CN(CCN1CCO)CCS(=O)(=O)O
C8H18N2O4S
MFCD00006158
23831
2-[4-(2-hydroxyethyl)piperazin-1-yl]ethanesulfonic acid
Safety and Handling

Safety and Handling

TSCA : TSCA

storageNote1 : 2° to 8°C

Documents