Please login to your online account to display your discounted pricing

Alfa Aesar™ HEPES Buffer, 0.2M buffer soln., pH 7.6, low endotoxin


Quantity 100mL

Manufacturer: alfa aesar™  J67738AE

 View more versions of this product

Catalog No. AAJ67738AE

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

CAS 7365-45-9
TSCA TSCA
Name Note low endotoxin
Physical Form Liquid
MDL Number MFCD00006158
Storage Note 1 2° to 8°C
Chemical Name or Material HEPES Buffer
pH 7.6
Color Colorless
Quantity 100mL
Concentration 0.2M buffer soln.

Specifications

Chemical Name or Material HEPES Buffer
CAS 7365-45-9
Name Note low endotoxin
MDL Number MFCD00006158
Accessories

2° to 8°C