Please login to your online account to display your discounted pricing

Alfa Aesar™ HEPES Buffer, 0.2M buffer soln., pH 7.6, low endotoxin

Chemical Identifiers

Chemical Name or Material HEPES Buffer
CAS 7365-45-9
Name Note low endotoxin
MDL Number MFCD00006158
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAJ67738AE

 
alfa aesar™
J67738AE
100mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAJ67738AK

 
alfa aesar™
J67738AK
250mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

CAS 7365-45-9
MDL Number MFCD00006158
TSCA TSCA
Physical Form Liquid
Storage Note 1 2° to 8°C
Name Note low endotoxin
Color Colorless
Concentration 0.2M buffer soln.
Quantity 100mL
pH 7.6
Chemical Name or Material HEPES Buffer
Safety and Handling

2° to 8°C