Please login to your online account to display your discounted pricing

Alfa Aesar™ HEPES Buffer, 0.2M buffer soln., pH 7.5, low endotoxin

Chemical Identifiers

Chemical Name or Material HEPES Buffer
CAS 7365-45-9
Name Note low endotoxin
MDL Number MFCD00006158
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAJ67257AE

 
alfa aesar™
J67257AE
100mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAJ67257AK

 
alfa aesar™
J67257AK
250mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Chemical Name or Material HEPES Buffer
pH 7.5
Color Colorless
Quantity 100mL
TSCA TSCA
Name Note low endotoxin
Physical Form Liquid
Concentration 0.2M buffer soln.
MDL Number MFCD00006158
CAS 7365-45-9
Storage Note 1 2° to 8°C
Safety and Handling

2° to 8°C