Please login to your online account to display your discounted pricing

Alfa Aesar™ HEPES Buffer, 0.2M buffer soln., pH 7.0, low endotoxin

Chemical Identifiers

Chemical Name or Material HEPES Buffer
CAS 7365-45-9
Name Note low endotoxin
MDL Number MFCD00006158
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAJ67405AE

 
alfa aesar™
J67405AE
100mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAJ67405AK

 
alfa aesar™
J67405AK
250mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Chemical Name or Material HEPES Buffer
TSCA TSCA
Name Note low endotoxin
Physical Form Liquid
Concentration 0.2M buffer soln.
pH 7.0
Color Colorless
Quantity 100mL
MDL Number MFCD00006158
CAS 7365-45-9
Storage Note 1 2° to 8°C

Applications
May be used in isoelectric focusing applications and as a buffer in the quantitative and selective measurement of antigen-antibody reactions.

Solubility
Soluble in water (1 M) at 20°C.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from strong oxidizing agents.

Safety and Handling

2° to 8°C