missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ HEPES, 0.5M buffer soln., pH 9.0

$72.93 - $109.03

Chemical Identifiers

CAS 7365-45-9
Molecular Formula C8H18N2O4S
Molecular Weight (g/mol) 238.302
MDL Number MFCD00006158
InChI Key JKMHFZQWWAIEOD-UHFFFAOYSA-N
PubChem CID 23831
ChEBI CHEBI:42334
IUPAC Name 2-[4-(2-hydroxyethyl)piperazin-1-yl]ethanesulfonic acid
SMILES C1CN(CCN1CCO)CCS(=O)(=O)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAJ63614AE View Documents Alfa Aesar™
J63614AE
100mL Each for $72.93
 
AAJ63614AK View Documents Alfa Aesar™
J63614AK
250mL Each for $109.03
 
Specifications

Chemical Identifiers

7365-45-9
238.302
JKMHFZQWWAIEOD-UHFFFAOYSA-N
CHEBI:42334
C1CN(CCN1CCO)CCS(=O)(=O)O
C8H18N2O4S
MFCD00006158
23831
2-[4-(2-hydroxyethyl)piperazin-1-yl]ethanesulfonic acid

Specifications

7365-45-9
MFCD00006158
JKMHFZQWWAIEOD-UHFFFAOYSA-N
2-[4-(2-hydroxyethyl)piperazin-1-yl]ethanesulfonic acid
23831
Liquid
100mL
C8H18N2O4S
0.5M buffer soln.
C1CN(CCN1CCO)CCS(=O)(=O)O
238.302
CHEBI:42334
9.0
HEPES buffer soln.
Safety and Handling

Safety and Handling

TSCA : TSCA