Please login to your online account to display your discounted pricing

Thermo Scientific™ Hepatocyte Growth Factor Ab-2, Mouse Monoclonal Antibody

Ideal for Western Blotting, Thermo Scientific™ Hepatocyte Growth Factor Ab-2, Mouse Monoclonal Antibody provides accurate, reproducible results.

$236.71 - $1,181.51

Specifications

Antigen Hepatocyte Growth Factor Ab-2
Clone SBF5
Classification Monoclonal
Conjugate Unlabeled
Cross Reactivity Human
View More Specs
Products
Catalog Number Mfr. No. Concentration Quantity Price Quantity    

MS775P

 
thermo scientific™
MS775P
200μg/mL 1mL Each for $1,096.96

MS775P0

 
thermo scientific™
MS775P0
200μg/mL 100μL Each for $236.71

MS775P1

 
thermo scientific™
MS775P1
200μg/mL 500μL Each for $695.96

MS775P1ABX

 
thermo scientific™
MS775P1ABX
1mg/mL 100μL Each for $774.37

MS775PABX

 
thermo scientific™
MS775PABX
1mg/mL 200μL Each for $1,181.51
Description & Specifications

Specifications

Antigen Hepatocyte Growth Factor Ab-2
Clone SBF5
Classification Monoclonal
Conjugate Unlabeled
Cross Reactivity Human
Format Concentrated
Host Species Murine
Regulatory Status RUO
Research Discipline Growth Factor
Type Primary
Immunogen Recombinant human HGF protein
Isotype IgG1

Ideal for Western Blotting, Thermo Scientific™ Hepatocyte Growth Factor Ab-2, Mouse Monoclonal Antibody provides accurate, reproducible results.

Hepatocyte Growth Factor (HGF), also known as Scatter Factor (SF) or Hepatopoietin A. Membrane receptor for HGF/SF is the product of the proto-oncogene c-met, a tyrosine kinase. HGF/SF is a pleiotropic growth factor which is produced by mesodermally derived cells, such as Kupffer cells/macrophages, endothelial cells, or hepatic fat-storing cells.

Host Species: Mouse

Clone: SBF5

Isotype: IgG1

Species Reactivity: Human. Others not known.

Epitope: Not determined

Immunogen: Recombinant human HGF protein

Molecular Weight: ∽82kDa

Recommended for:

  • Western Blotting (Not verified)

Note:

USA: RUO; Int'l: RUO