missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Thermo Scientific™ Hepatocyte Growth Factor Ab-1, Mouse Monoclonal Antibody

Ensure accurate, reproducible results with Thermo Scientific™ Hepatocyte Growth Factor Ab-1, Mouse Monoclonal Antibody.

$244.18 - $1,194.02

Specifications

Antigen Hepatocyte Growth Factor Ab-1
Clone 4C12.1
Classification Monoclonal
Conjugate Unlabeled
Format Concentrated
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Concentration Quantity Price Quantity  
Catalog Number Mfr. No. Concentration Quantity Price Quantity  
MS651P Thermo Scientific™
MS651P
200μg/mL 1mL Each for $1,132.96
MS651P0 Thermo Scientific™
MS651P0
200μg/mL 100μL Each for $244.18
MS651P1 Thermo Scientific™
MS651P1
200μg/mL 500μL Each for $709.90
MS651P1ABX Thermo Scientific™
MS651P1ABX
1mg/mL 100μL Each for $782.65
MS651PABX Thermo Scientific™
MS651PABX
1mg/mL 200μL Each for $1,194.02
Specifications

Description

Ensure accurate, reproducible results with Thermo Scientific™ Hepatocyte Growth Factor Ab-1, Mouse Monoclonal Antibody.

Hepatocyte Growth Factor (HGF), also known as Scatter Factor (SF) or Hepatopoietin A. Membrane receptor for HGF/SF is the product of the proto-oncogene c-met, a tyrosine kinase. HGF/SF is a pleiotropic growth factor which is produced by mesodermally derived cells, such as Kupffer cells/macrophages, endothelial cells, or hepatic fat-storing cells.

Host Species: Mouse

Clone: 4C12.1

Isotype: IgG2b

Species Reactivity: Human. Others not known.

Epitope: Not determined

Immunogen: Recombinant human HGF protein

Molecular Weight: ∽82kDa

Recommended for:

  • Western Blotting (Not verified)

Note:

USA: RUO; Int'l: RUO

Description & Specifications

Specifications

Hepatocyte Growth Factor Ab-1
Monoclonal
Concentrated
RUO
Primary
IgG2b
4C12.1
Unlabeled
Murine
Growth Factor
Recombinant human HGF protein