missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Heparin, Fisher BioReagents

Manufacturer:  Fisher Scientific Company BP2524100

 View more versions of this product

Catalog No. BP2524100



 

Specifications Description & Specifications

Specifications

Powder/Solid
100mg
White to Off-white
≤5.0%
Pass Test
Vapor Pressure: Negligible
≤0.03 EU/HEP
Report U/mg (Anti-XA)
9041-08-1
≈5000
15, 4688
5.0 to 7.5
0.003% max.
Poly Vial
≥140hep/mg (dry basis)
1.3 to 2.5%
Heparin
From Porcine Intestinal Mucosa

Chemical Identifiers

9041-08-1
≈5000
Heparin, From Porcine Intestinal Mucosa

Safety and Handling

SDS & Safety Documents

CAUTION!

Emergency Overview
Handle in accordance with good industrial hygiene and safety practice. The toxicological properties have not been fully investigated. Use personal protective equipment. Wash off immediately with plenty of water for at least 15 minutes. Get medical attention immediately if symptoms occur. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. If breathing is difficult, give oxygen. Get medical attention immediately if symptoms occur. Do not induce vomiting. Obtain medical attention. Obtain medical attention. .

NFPA

Health:1
Flammability:0
Instability:0

Recommended Storage : RT