missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Heparin, Fisher BioReagents

$189.30 - $189.30

Chemical Identifiers

CAS 9041-08-1
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
BP2524250 SDS View Documents Fisher BioReagents
BP2524250
250mg Poly Vial Each for $189.30
Case of 5 Each for $946.50
 
Description

Description

Heparin is used as an anticoagulant in biochemical research.
Specifications

Chemical Identifiers

9041-08-1

Specifications

Pass Test
Poly Vial
White
5.0 to 7.5
15, 4688
Powder/Solid
≤5.0%
Negligible
250mg
9041-08-1
≈5000
Heparin, From Porcine Intestinal Mucosa
Safety and Handling

Safety and Handling

CAUTION!

Emergency Overview
Handle in accordance with good industrial hygiene and safety practice. The toxicological properties have not been fully investigated. Use personal protective equipment. Wash off immediately with plenty of water for at least 15 minutes. Get medical attention immediately if symptoms occur. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. If breathing is difficult, give oxygen. Get medical attention immediately if symptoms occur. Do not induce vomiting. Obtain medical attention. Obtain medical attention. .

NFPA

Health:1
Flammability:0
Instability:0

Recommended Storage : RT