Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ Heparan Sulfate Proteoglycan/Perlecan Ab-1, Rat Monoclonal Antibody

Obtain accurate, reproducible results in immunohistochemistry and immunofluorescence experiments.

$280.59 - $1,523.53

Specifications

Antigen Heparan Sulfate Proteoglycan/Perlecan Ab-1
Clone A7L6
Applications Immunohistochemistry (Frozen)
Classification Monoclonal
Cross Reactivity Bovine
View More Specs
Products
Catalog Number Mfr. No. Concentration Conjugate Quantity Price Quantity    

RT794B

 
thermo scientific™ lab vision™
RT794B
200µg/mL Biotin 1mL Each for $1,523.53

RT794B0

 
thermo scientific™ lab vision™
RT794B0
200µg/mL Biotin 100µL Each for $374.86

RT794B1

 
thermo scientific™ lab vision™
RT794B1
200µg/mL Biotin 500µL Each for $1,020.16

RT794P

 
thermo scientific™ lab vision™
RT794P
200µg/mL Unlabeled 1mL Each for $1,195.33

RT794P0

 
thermo scientific™ lab vision™
RT794P0
200µg/mL Unlabeled 100µL Each for $280.59

RT794P1

 
thermo scientific™ lab vision™
RT794P1
200µg/mL Unlabeled 500µL Each for $778.98

RT794P1ABX

 
thermo scientific™ lab vision™
RT794P1ABX
1mg/mL Unlabeled 100µL Each for $861.92

RT794PABX

 
thermo scientific™ lab vision™
RT794PABX
1mg/mL Unlabeled 200µL Each for $1,282.55
Description & Specifications

Specifications

Antigen Heparan Sulfate Proteoglycan/Perlecan Ab-1
Clone A7L6
Applications Immunohistochemistry (Frozen)
Classification Monoclonal
Cross Reactivity Bovine
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Bovine
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Bovine
Cross Reactivity Human
Cross Reactivity Mouse
Format Concentrated
Host Species Rat
Regulatory Status RUO
Research Discipline Angiogenesis
Type Primary
Immunogen Murine EHS laminin preparation
Isotype IgG2a kappa

Obtain accurate, reproducible results in immunohistochemistry and immunofluorescence experiments with Thermo Scientific™ Heparan Sulfate Proteoglycan/Perlecan Ab-1, Rat Monoclonal Antibody.

Perlecan is a major heparan-sulfate proteoglycan (HSPG) found within all basement membranes and cell surfaces. Because of its strategic location and ability to store and protect growth factors, perlecan has been strongly implicated in the control of tumor cell growth and metastatic behavior. Perlecan possesses angiogenic and growth-promoting attributes primarily by acting as a coreceptor for basic fibroblast growth factor (FGF-2). Suppression of perlecan causes substantial inhibition of neoplastic growth and neovascularization. Thus, perlecan is a potent inducer of neoplasm growth and angiogenesis in vivo and therapeutic interventions targeting this key modulator of tumor progression may improve neoplastic treatment.

Host Species: Rat

Clone: A7L6

Isotype: IgG2a/κ

Species Reactivity: Human, Cow, Pig, Mouse. Others not known.

Epitope: Not determined

Immunogen: Murine EHS laminin preparation

Molecular Weight: High

Positive Control: Tonsil

Cellular Localization: Basement membrane

Recommended for:

  • Immunohistochemistry (Frozen and acid-alcohol-fixed, paraffin-embedded)
  • Western Blotting (Not verified)
  • Immunofluorescence

Note:

USA: RUO; Int'l: RUO