Thermo Scientific™ Replacement HEPA Filters and Cleaning Kits
Help ensure measurable Class 100 air quality performance with Thermo Scientific™ Replacement HEPA Filters and Cleaning Kits.
Manufacturer:  Thermo Scientific™ FOR1900160
 View more versions of this product
Catalog No. 15-467-654
Description
Description
Airborne particulates are a primary source of contamination in most lab settings.  Our advanced HEPA filter technology protects your cultures, providing ISO-Class 5 clean room-like air quality conditions within only five minutes after a 30-second door opening.
HEPA Filters
  • 99.97% filtering at 0.3μm
  • Compatible with all Thermo Scientific™ Forma™ Series II 3110, Thermo Scientific™ Steri-Cycle™, and Forma™ 310 Series (with HEPA filtration system option) incubators
  • Main chamber filter available as a stand-alone or packaged in a group with the other chamber in-line filters
Decontamination Kits
  • Consists of filters for difficult-to-clean chambers and gaskets (except door) and tubing
  • Reduces downtime and labor spent on decontamination
Specifications
Specifications
Steri-Cult 3307/3308, 3310/3311 incubators
HEPA Filter Replacement Kit
Documents