missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Heneicosafluorodecyl Iodide, TCI America™

$44.31 - $132.92

Chemical Identifiers

CAS 423-62-1
Molecular Formula C10F21I
Molecular Weight (g/mol) 645.981
MDL Number MFCD00001065
InChI Key UDWBMXSQHOHKOI-UHFFFAOYSA-N
Synonym perfluorodecyl iodide, 1-iodoperfluorodecane, perfluoro-1-iododecane, henicosafluoro-10-iododecane, heneicosafluorodecyl iodide, heneicosafluoro-1-iododecane, decane, heneicosafluoro-1-iodo, iodoperfluorodecane, henicosafluorodecyl iodide, 1-iodohenicosafluorodecane
PubChem CID 67920
IUPAC Name 1,1,1,2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10-henicosafluoro-10-iododecane
SMILES C(C(C(C(C(C(F)(F)I)(F)F)(F)F)(F)F)(F)F)(C(C(C(C(F)(F)F)(F)F)(F)F)(F)F)(F)F
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
H084425G View Documents TCI America
H084425G
25g Each for $132.92
 
H08445G View Documents TCI America
H08445G
5g Each for $44.31
 
Specifications

Chemical Identifiers

423-62-1
645.981
UDWBMXSQHOHKOI-UHFFFAOYSA-N
67920
C(C(C(C(C(C(F)(F)I)(F)F)(F)F)(F)F)(F)F)(C(C(C(C(F)(F)F)(F)F)(F)F)(F)F)(F)F
C10F21I
MFCD00001065
perfluorodecyl iodide, 1-iodoperfluorodecane, perfluoro-1-iododecane, henicosafluoro-10-iododecane, heneicosafluorodecyl iodide, heneicosafluoro-1-iododecane, decane, heneicosafluoro-1-iodo, iodoperfluorodecane, henicosafluorodecyl iodide, 1-iodohenicosafluorodecane
1,1,1,2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10-henicosafluoro-10-iododecane

Specifications

102°C
67°C
423-62-1
MFCD00001065
UDWBMXSQHOHKOI-UHFFFAOYSA-N
1,1,1,2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10-henicosafluoro-10-iododecane
67920
Crystal-Powder at 20°C
Heneicosafluorodecyl Iodide
Red-White
25g
C10F21I
perfluorodecyl iodide, 1-iodoperfluorodecane, perfluoro-1-iododecane, henicosafluoro-10-iododecane, heneicosafluorodecyl iodide, heneicosafluoro-1-iododecane, decane, heneicosafluoro-1-iodo, iodoperfluorodecane, henicosafluorodecyl iodide, 1-iodohenicosafluorodecane
C(C(C(C(C(C(F)(F)I)(F)F)(F)F)(F)F)(F)F)(C(C(C(C(F)(F)F)(F)F)(F)F)(F)F)(F)F
645.981
645.98
≥98.0% (GC)
Safety and Handling

Safety and Handling

EINECSNumber : (2)-0090

RTECSNumber : HD8710000

TSCA : Yes