missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Hemokinin 1, Human

For research use

$464.86 - $464.86

Specifications

Formulation Lyophilized powder
Recombinant No
Storage Requirements -30° to -10°C
Product Type Hemokinin 1, Human
Format Lyophilized Powder
View More Specs

Disclaimers:

For Research Use Only



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAJ65288LB0 Alfa Aesar™
J65288LB0
5mg N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AAJ65288MA Alfa Aesar™
J65288MA
10mg Each for $464.86
Specifications Description & Specifications

Specifications

Lyophilized powder
-30° to -10°C
Lyophilized Powder
No
Hemokinin 1, Human
RUO