Please login to your online account to display your discounted pricing

Alfa Aesar™ Hemoglobin Digest Standard, Certified Mass Spec Grade


Manufacturer: alfa aesar™  J67770LPL

 View more versions of this product

Catalog No. AAJ67770LPL

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Specifications

For Use With (Application) For the standardizing, calibrating, and troubleshooting chromatographic separations. Particularly useful for LC-MS applications.
CAS 9008-02-0
Quantity 1 vial
Product Type Hemoglobin Digest Standard
Solubility Soluble in water
Description Red
Storage Requirements -30°C to -10°C
Format Lyophilized powder