missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Hematoxylin Trihydrate, Fisher BioReagents

Manufacturer:  Fisher Scientific Company BP2523100

 View more versions of this product

Catalog No. BP2523100



 

Specifications Description & Specifications

Specifications

356.4
100°C
15, 4672
Amber
Amber Glass
Report %
290 to 292nm
Powder/Solid
100g
C16H14O6 · xH2O
Natural Black I
517-28-2
Pass Test
Hematoxylin Trihydrate

Chemical Identifiers

517-28-2
C16H14O6 · xH2O
Natural Black I
Powder/Solid
356.4
Hematoxylin Trihydrate

Safety and Handling

SDS & Safety Documents

Recommended Storage : RT